missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-1,2-Propane-1,1,2-d3-diamine, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 1,2-Diaminopropane-1,1,2-D3 |
| CAS | 2708283-46-7 |
| Molecular Formula | C3 D3 H7 N2 |
| Quantity | 0.05 g |
| Synonym | 1,2-Diaminopropane-1,1,2-D3, 1,1,2-Trideuteriopropane-1,2-diamine, 1,2-Propanediamine-1,1,2-d3, (±)-1,2-Diaminopropane-1,1,2-d3, 1-Methyl-1,2-diaminoethane-1,2,2-d3, Propylenediamine-d3 |
| InChI Formula | InChI=1 S/C3H10N2/c1-3(5)2-4/h3H,2,4-5H2,1H3/i2D2,3 D |
| SMILES | [2 H]C([2 H])(N)C([2 H])(C)N |
| IUPAC Name | 1,1,2-trideuteriopropane-1,2-diamine |
| Molecular Weight (g/mol) | 77.1434 |
| Formula Weight | 77.1032 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction