missing translation for 'onlineSavingsMsg'
Learn More

1-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]-1-propanone, TRC

Product Code. 30008024
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008024 100 mg 100mg
30005544 25 mg 25mg
30009009 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008024 Supplier Toronto Research Chemicals Supplier No. TRCC380065100MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1258428-71-5
Molecular Formula C29 H26 Cl N O2
Molecular Weight (g/mol) 455.98
Synonym 1-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]-1-propanone, Montelukast Impurity 1
IUPAC Name 1-[3-[(E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl]-3-[2-(2-hydroxypropan-2-yl)phenyl]propan-1-one
SMILES CC(C)(O)c1ccccc1CCC(=O)c2cccc(\C=C\c3ccc4ccc(Cl)cc4n3)c2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C29H26ClNO2/c1-29(2,33)26-9-4-3-7-21(26)13-17-28(32)23-8-5-6-20(18-23)10-15-25-16-12-22-11-14-24(30)19-27(22)31-25/h3-12,14-16,18-19,33H,13,17H2,1-2H3/b15-10+
Formula Weight 455.1652
Percent Purity >95%
Chemical Name or Material 1-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]-1-propanon
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.