missing translation for 'onlineSavingsMsg'
Learn More

1-(3,4-Dihydro-7-hydroxy-6-methoxy-2(1H)-isoquinolinyl)-1-propanone, TRC

Product Code. 30384161
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30384161 50 mg -
30369979 250 mg 250mg
30379690 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384161 Supplier Toronto Research Chemicals Supplier No. TRCD44499550MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1032822-42-6
Molecular Formula C13H17NO3
Molecular Weight (g/mol) 235.28
SMILES CCC(=O)N1CCc2cc(OC)c(O)cc2C1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H17NO3/c1-3-13(16)14-5-4-9-7-12(17-2)11(15)6-10(9)8-14/h6-7,15H,3-5,8H2,1-2H3
Formula Weight 235.1208
Chemical Name or Material 1-(3,4-Dihydro-7-hydroxy-6-methoxy-2(1H)-isoquinolinyl)-1-propanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.