missing translation for 'onlineSavingsMsg'
Learn More

1-(3,5-Dimethylphenyl)ethanone, 98%, BLD Pharm™

Product Code. 31007332
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
Unit Size:
100g
10g
1g
25g
500g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31007332 100 g 100g
31006842 1 g 1g
31000235 25 g 25g
30997454 500 g 500g
30990989 10 g 10g
30989937 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31007332 Supplier BLD Pharm Supplier No. BD12013100g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 5379-16-8
Molecular Formula C10H12O
MDL Number MFCD01075693
Quantity 100 g
SMILES CC(=O)C1=CC(C)=CC(C)=C1
Molecular Weight (g/mol) 148.20
Percent Purity 98%
Chemical Name or Material 1-(3,5-Dimethylphenyl)ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.