Learn More
1,3,5-Tri(1-naphthyl)benzene, TRC
Product Code. 30454607
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454607 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30454607 Supplier Toronto Research Chemicals Supplier No. TRCT88630810MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,3,5-Tri(1-naphthyl)benzene
CAS 7059-70-3
Molecular Formula C36H24
Quantity 10 mg
InChI Formula InChI=1S/C36H24/c1-4-16-31-25(10-1)13-7-19-34(31)28-22-29(35-20-8-14-26-11-2-5-17-32(26)35)24-30(23-28)36-21-9-15-27-12-3-6-18-33(27)36/h1-24H
SMILES c1ccc2c(cccc2c1)c3cc(cc(c3)c4cccc5ccccc45)c6cccc7ccccc67
IUPAC Name 1-(3,5-dinaphthalen-1-ylphenyl)naphthalene
Molecular Weight (g/mol) 456.58
Formula Weight 456.1878