missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tribenzoylbenzene, 96%, Thermo Scientific™

Product Code. 11438797
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11438797 5 g 5g
11428797 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11438797 Supplier Thermo Scientific Alfa Aesar Supplier No. A19941.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as a ligand in the synthesis and characterization of three new coordination polymers from the tritopic ligand.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25871-69-6
Molecular Formula C27H18O3
Molecular Weight (g/mol) 390.44
MDL Number MFCD00016577
InChI Key FDSRSUAVHPFWGT-UHFFFAOYSA-N
PubChem CID 141248
IUPAC Name (3,5-dibenzoylphenyl)-phenylmethanone
SMILES O=C(C1=CC=CC=C1)C1=CC(=CC(=C1)C(=O)C1=CC=CC=C1)C(=O)C1=CC=CC=C1

Specifications

Melting Point 114°C to 121°C
Quantity 5 g
Beilstein 1891480
Synonym 1,3,5-tribenzoylbenzene, benzene-1,3,5-triyltris phenylmethanone, 1,3,5-tribenzoyl benzene, methanone,1,3,5-benzenetriyltris phenyl, methanone, 1,3,5-benzenetriyltris phenyl, acmc-1ckhm, 3,5-dibenzoylphenyl-phenylmethanone, 3,5-dibenzoylphenyl phenyl methanone, 3,5-bis phenylcarbonyl phenyl phenyl ketone
Formula Weight 390.44
Percent Purity 96%
Chemical Name or Material 1,3,5-Tribenzoylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.