missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trichloro-2-nitrobenzene, TRC

Product Code. 30452927
Change view
Click to view available options
Quantity:
2.5 g
5 g
10 g
Unit Size:
10g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30452927 10 g 10g
30451579 5 g 5g
30450656 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30452927 Supplier Toronto Research Chemicals Supplier No. TRCT77492510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18708-70-8
Molecular Formula C6 H2 Cl3 N O2
Molecular Weight (g/mol) 226.44
Synonym Benzene, 1,3,5-trichloro-2-nitro-, 1,3,5-Trichloro-2-nitrobenzene, 2,4,6-Trichloro-1-nitrobenzene, 2,4,6-Trichloronitrobenzene, NSC 10244
IUPAC Name 1,3,5-trichloro-2-nitrobenzene
SMILES [O-][N+](=O)c1c(Cl)cc(Cl)cc1Cl

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H2Cl3NO2/c7-3-1-4(8)6(10(11)12)5(9)2-3/h1-2H
Formula Weight 224.9151
Chemical Name or Material 1,3,5-Trichloro-2-nitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.