missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trifluorobenzene, 98+%, ACROS Organics™

Product Code. 10337560 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
10mL; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10337560 10mL; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10337560 Supplier Thermo Scientific Acros Supplier No. 188430100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 372-38-3
CAS Min % 98.0
CAS Max % 100.0
Color Colorless
Density 1.27g/mL
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Molecular Formula C6H3F3
Linear Formula C6H3F3
Refractive Index 1.4130 to 1.4150 (20°C, 589nm)
MDL Number MFCD00000333
Quantity 10 mL
Beilstein 05, IV, 638
Synonym benzene, 1,3,5-trifluoro, sym-trifluorobenzene, unii-bn94c411f8, pubchem1069, 1,5-trifluorobenzene, acmc-209iru, 2,4,6-trifluorophenyl, benzene,3,5-trifluoro, 1,3,5-tri-fluorobenzene, 1,3,5-trifluoro-benzene
InChI Key JXUKFFRPLNTYIV-UHFFFAOYSA-N
SMILES C1=C(C=C(C=C1F)F)F
IUPAC Name 1,3,5-trifluorobenzene
Molecular Weight (g/mol) 132.085
PubChem CID 9745
Formula Weight 132.08
Percent Purity ≥98.0%
Physical Form Liquid
Chemical Name or Material 1, 3, 5-Trifluorobenzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.