missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(4-nitrophenyl)urea, TRC

Product Code. 30133978
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133978 10 g 10g
17942465 5 g 5g
17933765 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133978 Supplier Toronto Research Chemicals Supplier No. TRCB50850010G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 587-90-6
Molecular Formula C13 H10 N4 O5
Molecular Weight (g/mol) 302.24
Synonym Carbanilide, 4,4'-dinitro- (6CI,7CI,8CI), N,N'-Bis(4-nitrophenyl)urea, 1,3-Bis(4-nitrophenyl)urea, 1,3-Bis(p-nitrophenyl)urea, 1,3-Di(4-nitrophenyl)urea, 4,4'-Dinitrocarbanilide, 4,4'-Dinitrodiphenylurea, DNC, N,N'-Bis(p-nitrophenyl)urea, N,N'-Di(p-nitrophenyl)urea, NSC 101086
IUPAC Name 1,3-bis(4-nitrophenyl)urea
SMILES [O-][N+](=O)c1ccc(NC(=O)Nc2ccc(cc2)[N+](=O)[O-])cc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C13H10N4O5/c18-13(14-9-1-5-11(6-2-9)16(19)20)15-10-3-7-12(8-4-10)17(21)22/h1-8H,(H2,14,15,18)
Formula Weight 302.0651
Percent Purity >95%
Chemical Name or Material N,N'-Bis-(4-nitrophenyl)urea
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.