missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(4-piperidinyl)propane, 97+%

Product Code. 11305689
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11305689 100 g 100g
11395679 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11305689 Supplier Thermo Scientific Alfa Aesar Supplier No. L20442.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 16898-52-5 | C13H26N2 | 210.365 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16898-52-5
Molecular Formula C13H26N2
Molecular Weight (g/mol) 210.365
MDL Number MFCD00038013
InChI Key OXEZLYIDQPBCBB-UHFFFAOYSA-N
Synonym 1,3-bis 4-piperidyl propane, 4,4'-trimethylenedipiperidine, 1,3-di-4-piperidylpropane, piperidine, 4,4'-1,3-propanediyl bis, di-pip, 4-3-piperidin-4-yl propyl piperidine, 4-3-4-piperidyl propyl piperidine, unii-eu53ec219i, 1,3-di 4-piperidyl propane, 1,3-bis 4-piperidinyl propane
PubChem CID 85631
IUPAC Name 4-(3-piperidin-4-ylpropyl)piperidine
SMILES C1CNCCC1CCCC2CCNCC2

Specifications

Melting Point 63°C to 68°C
Boiling Point 327°C
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 111615
Sensitivity Hygroscopic
Formula Weight 210.36
Percent Purity ≥97%
Chemical Name or Material 1,3-Bis(4-piperidinyl)propane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.