missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(chloromethyl)tetramethyldisiloxane, 97%, Thermo Scientific™

Product Code. 11391577
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11391577 10 g 10g
11301587 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11391577 Supplier Thermo Scientific Alfa Aesar Supplier No. L04436.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1,3-Bis(chloromethyl)tetramethyldisiloxane is a useful bifunctional silylating agent.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2362-10-9
MDL Number MFCD00000877

Specifications

Boiling Point 92°C to 93°C (20mmHg)
Quantity 10 g
Molecular Formula C6H16Cl2OSi2
Beilstein 506631
Flash Point 73°C (163°F)
Synonym 1,3-bis chloromethyl-1,1,3,3-tetramethyldisiloxane, 1,3-bis chloromethyl tetramethyldisiloxane, bis chloromethyl tetramethyldisiloxane, disiloxane, 1,3-bis chloromethyl-1,1,3,3-tetramethyl, tetramethylbis chloromethyl disiloxane, tetramethyl-1,3-bis chloromethyl disiloxane, chloromethyl chloromethyl dimethylsilyl oxy dimethylsilane, disiloxane,3-bis chloromethyl-1,1,3,3-tetramethyl, pubchem13666, acmc-2097an
Solubility Information Not miscible or difficult to mix with water.
InChI Key NBGGEWGFZUDQKZ-UHFFFAOYSA-N
SMILES C[Si](C)(CCl)O[Si](C)(C)CCl
IUPAC Name chloromethyl-[chloromethyl(dimethyl)silyl]oxy-dimethylsilane
Molecular Weight (g/mol) 231.26
PubChem CID 75385
Formula Weight 231.27
Percent Purity 97%
Refractive Index 1.44
Density 1.05
Chemical Name or Material 1,3-Bis(chloromethyl)tetramethyldisiloxane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.