missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(phenylthio)propane, 98%, Thermo Scientific™

Product Code. 10063430 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10063430 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10063430 Supplier Thermo Scientific Acros Supplier No. 300250050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 28118-53-8
CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.1300g/mL
Boiling Point 264°C (12.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C15H16S2
Refractive Index 1.6271 to 1.6291
MDL Number MFCD00012310
Quantity 5 g
Specific Gravity 1.13
Synonym 1,3-bis phenylthio propane, 3-phenylsulfanyl propyl sulfanyl benzene, 3-phenylthiopropylthio benzene, 3-phenylthio propylthio benzene, 3-phenylsulfanyl propyl sulfanyl benzene #, benzene,1,1'-1,3-propanediylbis thio bis
InChI Key YNNYXLLXSONLCU-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)SCCCSC2=CC=CC=C2
IUPAC Name 3-phenylsulfanylpropylsulfanylbenzene
Molecular Weight (g/mol) 260.42
PubChem CID 576113
Formula Weight 260.42
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 3-Bis(phenylthio)propane, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.