Learn More
1,3-Cyclohexadiene, 96%, stab. with 0.1% BHT
Product Code. 11413997
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11413997 25 g 25g
11403997 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11413997 Supplier Thermo Scientific Alfa Aesar Supplier No. A19394.14
 Restricted Item
In Stock
Add to basket
missing translation for 'validMainframeMsg'
CAS: 592-57-4 | C6H8 | 80.13 g/mol
1,3-Cyclohexadiene is used as a hydrogen donor in transfer hydrogenation. It is used in the conversion to benzene. It is useful in the study of proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,3-Cyclohexadiene is used as a hydrogen donor in transfer hydrogenation. It is used in the conversion to benzene. It is useful in the study of proteomics research.
Solubility
Slightly miscible with methanol. Immiscible with water.
Notes
Incompatible with strong oxidizing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 592-57-4
Molecular Formula C6H8
Molecular Weight (g/mol) 80.13
MDL Number MFCD00001532
InChI Key MGNZXYYWBUKAII-UHFFFAOYSA-N
Synonym 1,3-cyclohexadiene, cyclohexadiene, 1,2-dihydrobenzene, unii-jv5w0eg5bp, jv5w0eg5bp, 3-cyclohexen-1,2-ylene, 1,3-cyclohexadien, 1,3 cyclohexadiene, 2,4-cyclohexadiene, cyclohexadiene-1,3
PubChem CID 11605
ChEBI CHEBI:37610
IUPAC Name cyclohexa-1,3-diene
SMILES C1CC=CC=C1
Specifications
Melting Point -89°C
Density 0.847
Boiling Point 79°C to 81°C
Flash Point −8°C (−144°F)
Odor Unpleasant
Refractive Index 1.474
Quantity 25 g
UN Number UN3295
Beilstein 506024
Solubility Information Slightly miscible with methanol. Immiscible with water.
Formula Weight 80.13
Percent Purity 96%
Chemical Name or Material 1,3-Cyclohexadiene, stabilized with 0.1% BHT