missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dibromopropane-1,1,3,3-d4, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1,3-Dibromopropane-1,1,3,3-D4 |
| CAS | 64528-94-5 |
| Molecular Formula | C3 D4 H2 Br2 |
| Quantity | 0.25 g |
| Synonym | Propane-1,1,3,3-d4, 1,3-dibromo- (9 CI), Propane-1,1,3,3-d4, 1,2-dibromo- (6 CI), 1,1,3,3-Tetradeutero-1,3-dibromopropane, 1,3-Dibromopropane-1,1,3,3-d4, 1,3-Dibromopropane-d4, 1,3-Dibromopropane-1,1,3,3-D4 |
| InChI Formula | InChI=1 S/C3H6Br2/c4-2-1-3-5/h1-3H2/i2D2,3D2 |
| SMILES | [2 H]C([2 H])(Br)CC([2 H])([2 H])Br |
| IUPAC Name | 1,3-dibromo-1,1,3,3-tetradeuteriopropane |
| Molecular Weight (g/mol) | 205.9124 |
| Formula Weight | 203.9087 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction