missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimesitylimidazolium chloride, 95%

Product Code. 11411820
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11411820 1 g 1g
11421820 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11411820 Supplier Thermo Scientific Alfa Aesar Supplier No. H27535.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 141556-45-8 | C21H25ClN2 | 340.90 g/mol

Used as a phosphine-free ligand in various metal-catalyzed coupling reactions, often with advantageous results in difficult cases. For use in the Pd-catalyzed cross-coupling of aryl Grignards with aryl chlorides (Kumada reaction). Many examples have been recorded of the use of NHC ligands in the Suzuki coupling reaction, for examples utilizing 1,3-dimesitylimidazol-2-ylidene, in the coupling of arylboronic acids with relatively unreactive aryl chlorides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a phosphine-free ligand in various metal-catalyzed coupling reactions, often with advantageous results in difficult cases. For use in the Pd-catalyzed cross-coupling of aryl Grignards with aryl chlorides (Kumada reaction). Many examples have been recorded of the use of NHC ligands in the Suzuki coupling reaction, for examples utilizing 1,3-dimesitylimidazol-2-ylidene, in the coupling of arylboronic acids with relatively unreactive aryl chlorides.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141556-45-8
Molecular Formula C21H25ClN2
Molecular Weight (g/mol) 340.90
MDL Number MFCD02684541
InChI Key OTOSIXGMLYKKOW-UHFFFAOYSA-M
Synonym 1,3-bis 2,4,6-trimethylphenyl imidazolium chloride, 1,3-dimesityl-1h-imidazol-3-ium chloride, 1,3-dimesitylimidazolium chloride, 1,3-bis mesityl imidazolium chloride, imes.hcl, 1,3-bis 2,4,6-trimethylphenyl imidazoliumchloride, 1,3-2,4,6-trimethylphenyl imidazolium chloride, 1,3-bis-2,4,6-trimethylphenyl-1h-imidazolium chloride, 1,3-bis-2,4,6-trimethyl-phenyl-3h-imidazol-1-ium chloride, 1,3-dihydro-1,3-dimesityl-2h-imidazol-2-ylidene monohydrochloride
PubChem CID 2734211
IUPAC Name 1,3-bis(2,4,6-trimethylphenyl)imidazol-1-ium;chloride
SMILES [Cl-].CC1=CC(C)=C(N2C=C[N+](=C2)C2=C(C)C=C(C)C=C2C)C(C)=C1

Specifications

Melting Point >300°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 340.89
Percent Purity 96%
Chemical Name or Material 1,3-Dimesitylimidazolium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.