missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dioleoyl-2-stearoylglycerol, TRC

Product Code. 30012535
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012535 100 mg 100mg
30012466 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012535 Supplier Toronto Research Chemicals Supplier No. TRCD484305100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2410-29-9
Molecular Formula C57H106O6
Molecular Weight (g/mol) 887.47
Synonym (9Z)-9-Octadecenoic Acid 1,1'-[2-[(1-Oxooctadecyl)oxy]-1,3-propanediyl] Ester, (Z)-9-Octadecenoic Acid 1,1'-[2-[(1-Oxooctadecyl)oxy]-1,3-propanediyl] Ester, 2-Stearo-1,3-diolein, 2-Stearoyldiolein, Glyceryl 1,3-Dioleate-2-stearate, Triglyceride OSO,sn, Triglyceride OStO,sn
SMILES CCCCCCCCCCCCCCCCCC(=O)OC(COC(=O)CCCCCCC\C=C/CCCCCCCC)COC(=O)CCCCCCC\C=C/CCCCCCCC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C57H106O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-55(58)61-52-54(63-57(60)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)53-62-56(59)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h25-26,28-29,54H,4-24,27,30-53H2,1-3H3/b28-25-,29-26-
Formula Weight 886.7989
Chemical Name or Material 1,3-Dioleoyl-2-stearoylglycerol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.