missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Ethoxy-4-methoxy-phenyl)-2-methanesulfonylethylamine (~10% inorganics), TRC

Product Code. 30382234
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382234 250 mg
30373556 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30382234 Supplier Toronto Research Chemicals Supplier No. TRCE891705250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 253168-94-4
Molecular Formula C12H19NO4S
Molecular Weight (g/mol) 273.35
Synonym 3-Ethoxy-4-methoxy-alpha-[(methylsulfonyl)methyl]benzenemethanamine, 1-(3-Ethoxy-4-methoxyphenyl)-2-methylsulfonylethylamine, Apremilast EP Impurity H
SMILES CCOc1cc(ccc1OC)C(N)CS(=O)(=O)C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H19NO4S/c1-4-17-12-7-9(5-6-11(12)16-2)10(13)8-18(3,14)15/h5-7,10H,4,8,13H2,1-3H3
Formula Weight 273.1035
Chemical Name or Material 1-(3-Ethoxy-4-methoxy-phenyl)-2-methanesulfonylethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.