missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Hydroxypropyl)theobromine, TRC

Product Code. 30137565
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137565 1 g 1g
30132914 500 mg 500mg
30005472 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137565 Supplier Toronto Research Chemicals Supplier No. TRCH9963701G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59413-14-8
Molecular Formula C10 H14 N4 O3
Molecular Weight (g/mol) 238.243
Synonym 3,7-Dihydro-1-(3-hydroxypropyl)-3,7-dimethyl-1H-purine-2,6-dione;
IUPAC Name 1-(3-hydroxypropyl)-3,7-dimethylpurine-2,6-dione
SMILES CN1C(=O)N(CCCO)C(=O)c2c1ncn2C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H14N4O3/c1-12-6-11-8-7(12)9(16)14(4-3-5-15)10(17)13(8)2/h6,15H,3-5H2,1-2H3
Formula Weight 238.107
Chemical Name or Material 1-(3-hydroxypropyl)-3,7-dimethylpurine-2,6-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.