missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Propane-d6-dithiol, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 1,3-Propanedithiol-D6 |
| CAS | 1219803-51-6 |
| Molecular Formula | C3 D6 H2 S2 |
| Quantity | 0.25 g |
| Synonym | 1,3-Propane-d6-dithiol, 1,3-Propane-1,1,2,2,3,3-d6-dithiol, 1,3-Dimercaptopropane-1,1,2,2,3,3-d6, 1,3-Propanedithiol-d6, 1,3-Propanedithiol-D6 |
| InChI Formula | InChI=1 S/C3H8S2/c4-2-1-3-5/h4-5 H,1-3H2/i1D2,2D2,3D2 |
| SMILES | [2 H]C([2 H])C([2 H])([2 H])C([2 H])([2 H])S |
| IUPAC Name | 1,1,2,2,3,3-hexadeuteriopropane-1,3-dithiol |
| Molecular Weight (g/mol) | 114.2626 |
| Formula Weight | 114.0444 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction