Learn More
Hamilton™ 1/4-28 Fittings and Washers (12 ga.) 10/pk
Product Code. 11522335
Change view
Click to view available options
Packaging Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity Unit Size
11522335 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11522335 Supplier Hamilton™ Supplier No. 88807
  • ONLINE EXCLUSIVE PRICE
    145.00€
    133.20€ / Pack of 10
    Save 11.80€
    8% Off
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Hamilton offers a diverse line of inert fittings fittings and connectors made from CTFE. The fittings are designed for use with industry standard luer and luer lock products like our syringes as well as threaded 1/4-28 and M6 flat bottom ports.
Specifications
Type 1/4-28 Fittings and Washers (12 ga.) 10/pk
Packaging Quantity 10/Pk.