missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Aminophenyl)ethanone hydrochloride, 98%, BLD Pharm™

Product Code. 30988474
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30988474 5 g 5g
30977229 25 g 25g
30988262 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30988474 Supplier BLD Pharm Supplier No. BD111975g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 41784-08-1
Molecular Formula C8H10ClNO
MDL Number MFCD00060232
Quantity 5 g
SMILES CC(C1=CC=C(N)C=C1)=O.[H]Cl
Molecular Weight (g/mol) 171.62
Percent Purity 98%
Chemical Name or Material 1-(4-Aminophenyl)ethanone hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.