missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(diphenylphosphino)butane, 97%

Product Code. 11424607
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424607 1 g 1g
11434607 5 g 5g
11444607 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424607 Supplier Thermo Scientific Alfa Aesar Supplier No. B21122.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7688-25-7 | C28H28P2 | 426.48 g/mol

1,4-Bis(diphenylphosphino)butane is used in Suzuki reaction. It plays an essential role as a ligand in palladium(0)-catalyzed acylcyanation of arylacetylenes and deprotection of allyloxycarbamates. It acts as a ligand for alkylations, isomerization reactions and in organometallic chemistry. Further, it is used in Heck reaction, Negishi coupling and Sonogashira coupling. In addition to this, it is useful in organometallic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Bis(diphenylphosphino)butane is used in Suzuki reaction. It plays an essential role as a ligand in palladium(0)-catalyzed acylcyanation of arylacetylenes and deprotection of allyloxycarbamates. It acts as a ligand for alkylations, isomerization reactions and in organometallic chemistry. Further, it is used in Heck reaction, Negishi coupling and Sonogashira coupling. In addition to this, it is useful in organometallic chemistry.

Solubility
Soluble in chloroform. Slightly soluble in water

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7688-25-7
Molecular Formula C28H28P2
Molecular Weight (g/mol) 426.48
MDL Number MFCD00003051
InChI Key BCJVBDBJSMFBRW-UHFFFAOYSA-N
Synonym 1,4-bis diphenylphosphino butane, dppb, phosphine, 1,4-butanediylbis diphenyl, 4-diphenylphosphanyl butyl diphenylphosphane, unii-35hp6ltd2d, tetramethylenebis diphenylphosphine, 35hp6ltd2d, 4-diphenylphosphanylbutyl diphenyl phosphane, 1,4-bis-diphenylphosphino butane
PubChem CID 82124
IUPAC Name 4-diphenylphosphanylbutyl(diphenyl)phosphane
SMILES C(CCP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 132°C to 136°C
Linear Formula (C6H5)2P(CH2)4P(C6H5)2
Quantity 1 g
Beilstein 706446
Sensitivity Air Sensitive
Solubility Information Soluble in chloroform. Slightly soluble in water
Formula Weight 426.48
Percent Purity 98%
Chemical Name or Material 1,4-Bis(diphenylphosphino)butane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.