missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Carboxyphenyl)-3-methyl-5-pyrazolone, TRC

Product Code. 30334556
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Product Code. Quantity unitSize
30334556 100 mg 100mg
30340080 50 mg 50mg
2 options
This item is not returnable. View return policy

Product Code. 30334556

Brand: Toronto Research Chemicals TRCB496043100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(4-Carboxyphenyl)-3-methyl-5-pyrazolone
CAS 60875-16-3
Molecular Formula C11H10N2O3
Quantity 100 mg
Synonym 4-(4,5-Dihydro-3-methyl-5-oxo-1H-pyrazol-1-yl)benzoic Acid, 1-(4-Carboxyphenyl)-3-methyl-2-pyrazolin-5-one, 1-(4-Carboxyphenyl)-3-methyl-2-pyrazolinone, 1-(p-Carboxyphenyl)-3-methyl-5-pyrazolone, 4-(3-Methyl-5-oxo-2-pyrazolin-1-yl)benzoic Acid, 4-(3-Methyl-5-oxo-4,5-dihydro-1H-pyrazol-1-yl)benzoic Acid, 4-(3-Methyl-5-oxopyrazol-1-yl)benzoic Acid
InChI Formula InChI=1S/C11H10N2O3/c1-7-6-10(14)13(12-7)9-4-2-8(3-5-9)11(15)16/h2-5H,6H2,1H3,(H,15,16)
SMILES Cc1cc(O)n(n1)c2ccc(cc2)C(=O)O
IUPAC Name 4-(3-methyl-5-oxo-4H-pyrazol-1-yl)benzoic acid
Molecular Weight (g/mol) 218.21
Formula Weight 218.0691
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.