missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorobenzyl)piperazine 98%, ACROS Organics™

Product Code. 10288373 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10288373 1 g Glass bottle 1g
10444515 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10288373 Supplier Thermo Scientific Acros Supplier No. 391950010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 23145-88-2
Molecular Formula C11H15ClN2
Molecular Weight (g/mol) 210.705
MDL Number MFCD00040791
InChI Key GSJXJZOWHSTWOX-UHFFFAOYSA-N
Synonym 1-4-chlorobenzyl piperazine, 1-4-chlorophenyl methyl piperazine, 4-chlorobenzylpiperazine, 1-4-chlorobenzyl-piperazine, 1-4-chloro-benzyl-piperazine, 1-4-chloro benzyl piperazine, n-4-chlorobenzyl piperazine, 4-chlorophenyl methyl piperazine, norhomochlorcyclizine, chlorobenzylpiperazin
PubChem CID 134826
IUPAC Name 1-[(4-chlorophenyl)methyl]piperazine
SMILES C1CN(CCN1)CC2=CC=C(C=C2)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.1400g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5560 to 1.5580 (20°C, 589nm)
Quantity 1 g
Formula Weight 210.71
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 1-(4-Chlorobenzyl)piperazine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.