missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)-1-cyclopentanecarboxylic acid, 98%, Thermo Scientific™

Product Code. p-3736008 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10235830

Brand: Thermo Scientific Acros 161570050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 80789-69-1
Molecular Formula C12H13ClO2
Molecular Weight (g/mol) 224.69
MDL Number MFCD00001373
InChI Key QJNFJEMGWIQMJT-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl cyclopentanecarboxylic acid, 1-4-chlorophenyl-1-cyclopentanecarboxylic acid, 1-4-chlorophenyl cyclopentane-1-carboxylic acid, cyclopentanecarboxylic acid, 1-4-chlorophenyl, asischem d50961, timtec-bb sbb000649, acmc-209pk2, labotest-bb lt00453384, 1-4-chloro phenyl cyclopentanecarboxylic acid, 1-4-chloro-phenyl-cyclopentylcarboxylic acid
PubChem CID 97447
IUPAC Name 1-(4-chlorophenyl)cyclopentane-1-carboxylic acid
SMILES C1CCC(C1)(C2=CC=C(C=C2)Cl)C(=O)O

Specifications

Melting Point 160.0°C to 164.0°C
CAS Min % 97.5
Color Cream to Gray
CAS Max % 100.0
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder or Crystals
Assay Percent Range 98%
Linear Formula ClC6H4C5H8CO2H
Formula Weight 224.69
Percent Purity 98%
Chemical Name or Material 1-(4-Chlorophenyl)-1-cyclopentanecarboxylic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.