missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)biguanide Hydrochloride, TRC

Product Code. 17971355
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17971355 10 g 10g
17950655 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17971355 Supplier Toronto Research Chemicals Supplier No. TRCC37854010G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4022-81-5
Molecular Formula C8 H10 Cl N5 . Cl H
Molecular Weight (g/mol) 248.11
Synonym Imidodicarbonimidic diamide, N-(4-chlorophenyl)-, hydrochloride (1:1), Biguanide, 1-(p-chlorophenyl)-, monohydrochloride (8CI), Imidodicarbonimidic diamide, N-(4-chlorophenyl)-, monohydrochloride (9CI), 1-(p-Chlorophenyl)biguanide hydrochloride, 1-(p-Chlorophenyl)biguanide monohydrochloride, Aminylium, [amino[[[(4-chlorophenyl)amino]iminomethyl]amino]methylene]-, chloride, Aminylium, [amino[[[(4-chlorophenyl)amino]iminomethyl]amino]methylene]-, chloride (1:1), N-(4-Chlorophenyl)imidodicarbonimidic diamide monohydrochloride, N-p-Chlorophenylbiguanide hydrochloride
IUPAC Name 1-carbamimidoyl-3-(4-chlorophenyl)guanidine;hydrochloride
SMILES Cl.NC(=N)NC(=N)Nc1ccc(Cl)cc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H10ClN5.ClH/c9-5-1-3-6(4-2-5)13-8(12)14-7(10)11;/h1-4H,(H6,10,11,12,13,14);1H
Formula Weight 247.0392
Chemical Name or Material 1-(4-Chlorophenyl)biguanide Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.