missing translation for 'onlineSavingsMsg'
Learn More

1-[(4-Chlorophenyl)phenylmethyl]piperazine Hydrochloride, TRC

Product Code. 30133982
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133982 250 mg 250mg
30132787 100 mg 100mg
30012115 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133982 Supplier Toronto Research Chemicals Supplier No. TRCC274840250MG
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18719-22-7
Molecular Formula C17H20Cl2N2 • xHCl
Molecular Weight (g/mol) 323.26
Synonym Norchlorcyclizine Hydrochloride
IUPAC Name 1-((4-chlorophenyl)(phenyl)methyl)piperazine hydrochloride
SMILES ClC1=CC=C(C(N2CCNCC2)C3=CC=CC=C3)C=C1.[•HCl]

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C17H19ClN2.ClH/c18-16-8-6-15(7-9-16)17(14-4-2-1-3-5-14)20-12-10-19-11-13-20;/h1-9,17,19H,10-13H2;1H
Chemical Name or Material 1-[(4-Chlorophenyl)phenylmethyl]piperazine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.