missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexanedimethanol Diglycidyl Ether, TRC

Product Code. 30135214
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135214 10 g 10g
30133575 5 g 5g
30010001 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135214 Supplier Toronto Research Chemicals Supplier No. TRCC99797310G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14228-73-0
Molecular Formula C14H24O4
Molecular Weight (g/mol) 256.34
Synonym 2,2'-[1,4-Cyclohexanediylbis(methyleneoxymethylene)]bis[oxirane], 1,4-Bis(glycidoxymethyl)cyclohexane, 1,4-Bis(hydroxymethyl)cyclohexane diglycidyl Ether, 1,4-Bis[(2,3-epoxypropoxy)methyl]cyclohexane, 1,4-Bis[(glycidyloxy)methyl]cyclohexane, 1,4-Cyclohexanedimethanol diglycidyl Ether
IUPAC Name 2-[[4-(oxiran-2-ylmethoxymethyl)cyclohexyl]methoxymethyl]oxirane
SMILES C(OCC1CO1)C2CCC(COCC3CO3)CC2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C14H24O4/c1-2-12(6-16-8-14-10-18-14)4-3-11(1)5-15-7-13-9-17-13/h11-14H,1-10H2
Formula Weight 256.1675
Chemical Name or Material 1,4-Cyclohexanedimethanol Diglycidyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.