missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Cyclohexanediol (Cis/Trans Mixture), TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | 1,4-Cyclohexanediol (Cis/Trans Mixture) |
| CAS | 556-48-9 |
| Molecular Formula | C6H12O2 |
| Quantity | 50 g |
| Synonym | 1,4-Dihydroxycyclohexane, NSC 5651, Quinitol |
| InChI Formula | InChI=1S/C6H12O2/c7-5-1-2-6(8)4-3-5/h5-8H,1-4H2 |
| SMILES | OC1CCC(O)CC1 |
| IUPAC Name | cyclohexane-1,4-diol |
| Molecular Weight (g/mol) | 116.16 |
| Formula Weight | 116.0837 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction