missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diamino-2,3-dihydroanthraquinone, TRC

Product Code. 30360587
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30360587 10 g 10g
30353687 1 g 1g
30353688 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30360587 Supplier Toronto Research Chemicals Supplier No. TRCD41628010G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-63-0
Molecular Formula C14 H12 N2 O2
Molecular Weight (g/mol) 240.26
Synonym 1,4-Diamino-2,3-dihydro-9,10-anthracenedione, 2,3-Dihydro-1,4-diamino
IUPAC Name 1,4-diamino-2,3-dihydroanthracene-9,10-dione
SMILES NC1=C2C(=O)c3ccccc3C(=O)C2=C(N)CC1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C14H12N2O2/c15-9-5-6-10(16)12-11(9)13(17)7-3-1-2-4-8(7)14(12)18/h1-4H,5-6,15-16H2
Formula Weight 240.0899
Chemical Name or Material 1,4-Diamino-2,3-dihydroanthraquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.