missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dibromobutane-d8, CDN
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| Chemical Name or Material | 1,4-Dibromobutane-d8 |
| CAS | 68375-92-8 |
| Molecular Formula | C4 2H8 Br2 |
| Quantity | 5 g |
| InChI Formula | InChI=1 S/C4H8Br2/c5-3-1-2-4-6/h1-4H2/i1D2,2D2,3D2,4D2 |
| SMILES | [2 H]C([2 H])(Br)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])Br |
| IUPAC Name | 1,4-dibromo-1,1,2,2,3,3,4,4-octadeuteriobutane |
| Molecular Weight (g/mol) | 223.96 |
| Formula Weight | 221.9495 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction