missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dimethyl-1,2,3,4-tetrahydroquinoxaline-6-carbonitrile, 97%, Thermo Scientific™

Product Code. 11330607
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11330607 5 g 5g
10757095 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11330607 Supplier Thermo Scientific Maybridge Supplier No. CC37116DE
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 857283-87-5
Molecular Formula C11H13N3
Molecular Weight (g/mol) 187.246
MDL Number MFCD09702356
InChI Key NBANRNWRIHAGBJ-UHFFFAOYSA-N
Synonym 1,4-dimethyl-1,2,3,4-tetrahydroquinoxaline-6-carbonitrile
PubChem CID 24229481
IUPAC Name 1,4-dimethyl-2,3-dihydroquinoxaline-6-carbonitrile
SMILES CN1CCN(C2=C1C=CC(=C2)C#N)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 187.24
Percent Purity 97%
Chemical Name or Material 1,4-Dimethyl-1,2,3,4-tetrahydroquinoxaline-6-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.