missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane, TRC

Product Code. 18001420
Change view
Click to view available options
Quantity:
1 g
100 g
Unit Size:
100g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18001420 100 g 100g
18091320 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 18001420 Supplier Toronto Research Chemicals Supplier No. TRCD485880100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-91-1
Molecular Formula C4 H8 O2
Molecular Weight (g/mol) 88.11
Synonym 1,4-Dioxane, p-Dioxane (8CI), 1,4-Diethylene dioxide, 1,4-Dioxacyclohexane, 1,4-Dioxan, 1,4-Dioxin, tetrahydro-, Diethylene dioxide, Diethylene ether, Diethylene oxide, Dioxan, Dioxane, Dioxyethylene ether, NE 220, NSC 8728, p-Dioxan
IUPAC Name 1,4-dioxane
SMILES C1COCCO1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2
Formula Weight 88.0524
Chemical Name or Material 1,4-Dioxane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.