Learn More
1,4-Dithiane-2,5-diol, TRC
Product Code. 30003295
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
25 g
missing translation for 'unitSize':
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30003295 1 g 1g
30135734 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30003295 Supplier Toronto Research Chemicals Supplier No. TRCD4938401G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 40018-26-6
Molecular Formula C4 H8 O2 S2
Molecular Weight (g/mol) 152.24
Synonym 1,4-Dithian-2,5-diol, 2,5-Dihydroxy-1,4-dithiane, Mercaptoacetaldehyde Dimer, NSC 106280, p-Dithiane-2,5-diol;
IUPAC Name 1,4-dithiane-2,5-diol
SMILES OC1CSC(O)CS1
Specifications
Quantity 1 g
InChI Formula InChI=1S/C4H8O2S2/c5-3-1-7-4(6)2-8-3/h3-6H,1-2H2
Formula Weight 151.9966
Chemical Name or Material 1,4-Dithiane-2,5-diol