missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Fluorophenyl)cyclopentanecarbonitrile, 99%, Thermo Scientific™

Product Code. 10309270 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10309270 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10309270 Supplier Thermo Scientific Acros Supplier No. 304830050
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 83706-50-7
CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C12H12FN
Refractive Index 1.5140 to 1.516
MDL Number MFCD00800616
Quantity 5 g
Synonym 1-4-fluorophenyl cyclopentanecarbonitrile, 1-4-fluorophenyl cyclopentane-1-carbonitrile, 1-4-fluoro-phenyl-cyclopentanecarbonitrile, cyclopentanecarbonitrile,1-4-fluorophenyl
InChI Key MAGOCEYAQAZRNE-UHFFFAOYSA-N
SMILES C1CCC(C1)(C#N)C2=CC=C(C=C2)F
IUPAC Name 1-(4-fluorophenyl)cyclopentane-1-carbonitrile
Molecular Weight (g/mol) 189.09
PubChem CID 2733239
Formula Weight 189.09
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-(4-Fluorophenyl)cyclopentanecarbonitrile, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.