missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Methoxyphenyl)-7-oxo-6-[4-(2-oxopiperidin-1-yl)phenyl]-4,5,6,7-tetrahydro-1H-pyrazolo[3,4-c]pyridine-3-carboxylic Acid, TRC

Product Code. 17991605
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17991605 5 mg 5mg
17910805 2.5 mg 2.5mg
17900935 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17991605 Supplier Toronto Research Chemicals Supplier No. TRCM2664205MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 503614-92-4
Molecular Formula C25 H24 N4 O5
Molecular Weight (g/mol) 460.48
Synonym 1H-Pyrazolo[3,4-c]pyridine-3-carboxylic acid, 4,5,6,7-tetrahydro-1-(4-methoxyphenyl)-7-oxo-6-[4-(2-oxo-1-piperidinyl)phenyl]-, 4,5,6,7-Tetrahydro-1-(4-methoxyphenyl)-7-oxo-6-[4-(2-oxo-1-piperidinyl)phenyl]-1H-pyrazolo[3,4-c]pyridine-3-carboxylic acid, 1-(4-Methoxyphenyl)-7-oxo-6-[4-(2-oxopiperidin-1-yl)phenyl]-4,5,6,7-tetrahydro-1H-pyrazolo[3,4-c]pyridine-3-carboxylic acid
IUPAC Name 1-(4-methoxyphenyl)-7-oxo-6-[4-(2-oxopiperidin-1-yl)phenyl]-4,5-dihydropyrazolo[3,4-c]pyridine-3-carboxylic acid
SMILES COc1ccc(cc1)n2nc(C(=O)O)c3CCN(C(=O)c23)c4ccc(cc4)N5CCCCC5=O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C25H24N4O5/c1-34-19-11-9-18(10-12-19)29-23-20(22(26-29)25(32)33)13-15-28(24(23)31)17-7-5-16(6-8-17)27-14-3-2-4-21(27)30/h5-12H,2-4,13-15H2,1H3,(H,32,33)
Formula Weight 460.1747
Percent Purity >95%
Chemical Name or Material Apixaban acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.