missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Methoxyphenyl)propan-2-one, Mikromol™
Description
1-(4-Methoxyphenyl)propan-2-one
Specifications
Specifications
| Chemical Name or Material | 1-(4-Methoxyphenyl)propan-2-one |
| CAS | 122-84-9 |
| Analyte or Component Names | 1-(4-Methoxyphenyl)propan-2-one |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H12 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1-(4-Methoxyphenyl)-2-propanone, 2-Propanone, (p-methoxyphenyl)- (6CI,7CI), 2-Propanone, 1-(p-methoxyphenyl)- (8CI), 1-(4-Methoxyphenyl)-2-propanone, 1-(4-Methoxyphenyl)acetone, 1-(p-Anisyl)-2-propanone, 1-(p-Methoxyphenyl)-2-propanone, 4-Methoxybenzyl methyl ketone, 4-Methoxyphenylacetone, 4'-Methoxyphenyl-2-propanone, Anisketone, Anisyl methyl ketone, NSC 22983, NSC 46101, p-Acetonylanisole, p-Anisylacetone, p-Methoxybenzyl methyl ketone, p-Methoxyphenylacetone, Mebeverine Hydrochloride Impurity A, Mebeverine Impurity A |
| InChI Formula | InChI=1S/C10H12O2/c1-8(11)7-9-3-5-10(12-2)6-4-9/h3-6H,7H2,1-2H3 |
| SMILES | COc1ccc(CC(=O)C)cc1 |
| IUPAC Name | 1-(4-methoxyphenyl)propan-2-one |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?