missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Nitrophenyl)piperidine, 97%, Thermo Scientific™

Product Code. 11494317
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494317 25 g 25g
11484317 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11494317 Supplier Thermo Scientific Alfa Aesar Supplier No. B20399.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6574-15-8
Molecular Formula C11H14N2O2
Molecular Weight (g/mol) 206.245
MDL Number MFCD00023662
InChI Key SGPLAXFUDTWHRS-UHFFFAOYSA-N
Synonym 1-4-nitrophenyl piperidine, 1-p-nitrophenyl piperidine, n-4-nitrophenyl piperidine, 1-4-nitro-phenyl-piperidine, piperidine, 1-4-nitrophenyl, n-p-nitrophenyl piperidine, 4-nitrophenyl piperidine, pubchem9438, maybridge1_000682, acmc-20a12d
PubChem CID 81049
IUPAC Name 1-(4-nitrophenyl)piperidine
SMILES C1CCN(CC1)C2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 102°C to 105°C
Quantity 25 g
Formula Weight 206.24
Percent Purity 97%
Chemical Name or Material 1-(4-Nitrophenyl)piperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.