missing translation for 'onlineSavingsMsg'
Learn More

1,4-Piperazinediethylamine, TRC

Product Code. 30180361
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180361 1 g 1g
30180148 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30180361 Supplier Toronto Research Chemicals Supplier No. TRCP4800901G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6531-38-0
Molecular Formula C8H20N4
Molecular Weight (g/mol) 172.27
Synonym 1,4-Bis(2-aminoethyl)-1,4-diazacyclohexane, 1,4-Bis(2-aminoethyl)piperazine, N,N'-Bis(2-aminoethyl)piperazine, [2-[4-(2-Aminoethyl)piperazin-1-yl]ethyl]amine, 1,4-Piperazinediethanamine
IUPAC Name 2-[4-(2-aminoethyl)piperazin-1-yl]ethanamine
SMILES NCCN1CCN(CCN)CC1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H20N4/c9-1-3-11-5-7-12(4-2-10)8-6-11/h1-10H2
Formula Weight 172.1688
Chemical Name or Material 1,4-Piperazinediethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.