Learn More
1-(5,6,7,8-Tetrahydronaphthalen-2-yl)ethanone, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 774-55-0 |
| Molecular Formula | C12H14O |
| MDL Number | MFCD00019710 |
| Quantity | 25 g |
| SMILES | CC(C1=CC=C2CCCCC2=C1)=O |
| Molecular Weight (g/mol) | 174.24 |
| Percent Purity | 98% |
| Chemical Name or Material | 1-(5,6,7,8-Tetrahydronaphthalen-2-yl)ethanone |