Learn More
1,5-Dinitronaphthalene, 97+%
Product Code. 11449060
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449060 25 g 25g
11459060 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11449060 Supplier Thermo Scientific Alfa Aesar Supplier No. A13869.14
 Restricted Item
In Stock
Add to basket
CAS: 605-71-0 | C10H6N2O4 | 218.168 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 605-71-0
Molecular Formula C10H6N2O4
Molecular Weight (g/mol) 218.168
MDL Number MFCD00003916
InChI Key ZUTCJXFCHHDFJS-UHFFFAOYSA-N
Synonym naphthalene, 1,5-dinitro, naphthalene, dinitro, unii-ui2s14iw4t, ccris 6028, ui2s14iw4t, napththalene, 1,5-dinitro, 1,5 or 1,8-dinitronaphthalene, acmc-1bixi, 1,5-dinitro naphthalene, 1,5-dinitro-naphthalene
PubChem CID 11803
ChEBI CHEBI:50640
IUPAC Name 1,5-dinitronaphthalene
SMILES C1=CC2=C(C=CC=C2[N+](=O)[O-])C(=C1)[N+](=O)[O-]
Specifications
Melting Point 214°C to 218°C
Density 1.586
Odor Odorless
Quantity 25 g
Beilstein 527184
Formula Weight 218.17
Percent Purity ≥97%
Chemical Name or Material 1,5-Dinitronaphthalene