missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diphenyl-3-methylpyrazole, TRC

Product Code. 30388759
Change view
Click to view available options
Quantity:
25 mg
50 mg
200 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388759 200 mg
30376692 25 mg
30385638 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388759 Supplier Toronto Research Chemicals Supplier No. TRCD494883200MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3729-90-6
Molecular Formula C16H14N2
Molecular Weight (g/mol) 234.3
SMILES Cc1cc(c2ccccc2)n(n1)c3ccccc3

Specifications

Quantity 200 mg
InChI Formula InChI=1S/C16H14N2/c1-13-12-16(14-8-4-2-5-9-14)18(17-13)15-10-6-3-7-11-15/h2-12H,1H3
Formula Weight 234.1157
Chemical Name or Material 1,5-Diphenyl-3-methylpyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.