missing translation for 'onlineSavingsMsg'
Learn More

1,5-DipHenylcarbazone, Extra Pure, SLR, Fisher Chemical

Product Code. 10570911 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass Jar
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10570911 10 g Glass Jar 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10570911 Supplier Fisher Chemical Supplier No. D/4850/45

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 538-62-5
Melting Point 153°C
Color Orange
Packaging Glass Jar
Molecular Formula C13H12N4O
MDL Number 3024
Quantity 10 g
Synonym 1,5-diphenylcarbazone, diazenecarboxylic acid, phenyl-, 2-phenylhydrazide, unii-5uyj4r0d5t, phenylazoformic acid 2-phenylhydrazide, sym-diphenylcarbazone, 5uyj4r0d5t, chembl79409, 3-hydroxy-1,5-diphenylformazan, phenyldiazenecarboxylic acid 2-phenylhydrazide, 3-phenylamino-1-phenylimino urea
InChI Key ZFWAHZCOKGWUIT-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)NNC(=O)N=NC2=CC=CC=C2
IUPAC Name 1-anilino-3-phenyliminourea
Molecular Weight (g/mol) 240.266
PubChem CID 10860
Formula Weight 240.26g/mol
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.