missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,6-Dibromohexane-d12, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 1,6-Dibromohexane-D12 |
| CAS | 169397-96-0 |
| Molecular Formula | C6 D12 Br2 |
| Quantity | 0.1 g |
| Synonym | Hexane-1,1,2,2,3,3,4,4,5,5,6,6-d12, 1,6-dibromo- (9 CI), 1,6-Dibromohexane-d12, 1,6-Dibromohexane-D12, 1,6-Dibromohexane-1,1,2,2,3,3,4,4,5,5,6,6-d12, Hexamethylene-d12 dibromide |
| InChI Formula | InChI=1 S/C6H12Br2/c7-5-3-1-2-4-6-8/h1-6H2/i1D2,2D2,3D2,4D2,5D2,6D2 |
| SMILES | [2 H]C([2 H])(Br)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])Br |
| IUPAC Name | 1,6-dibromo-1,1,2,2,3,3,4,4,5,5,6,6-dodecadeuteriohexane |
| Molecular Weight (g/mol) | 256.0414 |
| Formula Weight | 254.0059 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction