missing translation for 'onlineSavingsMsg'
Learn More

1,6-Dibromohexane, TRC

Product Code. 30373185
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30373185 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30373185 Supplier Toronto Research Chemicals Supplier No. TRCD425585100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,6-Dibromohexane
CAS 629-03-8
Molecular Formula C6 H12 Br2
Quantity 100 g
Synonym Hexane, 1,6-dibromo-, 1,6-Dibromohexane, 1,6-Dibromo-n-hexane, Hexamethylene dibromide, NSC 7306, α,ω-Dibromohexane
InChI Formula InChI=1S/C6H12Br2/c7-5-3-1-2-4-6-8/h1-6H2
SMILES BrCCCCCCBr
IUPAC Name 1,6-dibromohexane
Molecular Weight (g/mol) 243.967
Formula Weight 241.9306
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.