missing translation for 'onlineSavingsMsg'
Learn More

1,6-Hexanedithiol, 97%

Product Code. 11326956
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11326956 5 g 5g
11336956 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11326956 Supplier Thermo Scientific Alfa Aesar Supplier No. L06686.06
 Restricted Item
Estimated Shipment: 17-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 1191-43-1 | C6H14S2 | 150.30 g/mol

1,6-Hexanedithiol is used in self-assembled monolayers (SAMs). It is used in synthetic rubber and food with spices. It is also used in Food Flavor, Tobacco Flavor, Blending edible flavors: Beef, poultry soup .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,6-Hexanedithiol is used in self-assembled monolayers (SAMs). It is used in synthetic rubber and food with spices. It is also used in Food Flavor, Tobacco Flavor, Blending edible flavors: Beef, poultry soup .

Solubility
Not miscible in water.

Notes
Incompatible materials are Bases, Oxidizing agents, Reducing agents, Alkali metals. Avoid Heat and direct sunlight.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1191-43-1
Molecular Formula C6H14S2
Molecular Weight (g/mol) 150.30
MDL Number MFCD00004910
InChI Key SRZXCOWFGPICGA-UHFFFAOYSA-N
Synonym 1,6-hexanedithiol, 1,6-dimercaptohexane, 1,6-hexanedimercaptan, usaf uctl-72, hexamethylene dimercaptan, 1,6-hexane dithiol, 1,6-hexamethylenedithiol, unii-rwn7rm884e, fema no. 3495, hexanedithiol-1,6
PubChem CID 14491
IUPAC Name hexane-1,6-dithiol
SMILES SCCCCCCS

Specifications

Melting Point -21°C
Density 0.99
Boiling Point 242°C to 243°C
Flash Point 90°C (194°F)
Refractive Index 1.513
Quantity 5 g
UN Number UN2810
Beilstein 1732507
Solubility Information Not miscible in water.
Formula Weight 150.3
Percent Purity 97%
Chemical Name or Material 1,6-Hexanedithiol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.