missing translation for 'onlineSavingsMsg'
Learn More

1-(6-Methyl-2-pyridinyl)-1H-imidazole-4-carboxylic Acid, TRC

Product Code. 30475133
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30475133 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30475133 Supplier Toronto Research Chemicals Supplier No. TRCM64157350MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(6-Methyl-2-pyridinyl)-1H-imidazole-4-carboxylic Acid
CAS 477847-81-7
Molecular Formula C10H9N3O2
Quantity 50 mg
InChI Formula InChI=1S/C10H9N3O2/c1-7-3-2-4-9(12-7)13-5-8(10(14)15)11-6-13/h2-6H,1H3,(H,14,15)
SMILES Cc1cccc(n1)n2cnc(c2)C(=O)O
IUPAC Name 1-(6-methylpyridin-2-yl)imidazole-4-carboxylic acid
Molecular Weight (g/mol) 203.19
Formula Weight 203.0695
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.