missing translation for 'onlineSavingsMsg'
Learn More

1,7-Dimethylfluorene, TRC

Product Code. 30011020
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011020 50 mg 50mg
30012102 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011020 Supplier Toronto Research Chemicals Supplier No. TRCD46573050MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 442-66-0
Molecular Formula C15 H14
Molecular Weight (g/mol) 194.27
Synonym 1,7-dimethyl-9H-fluorene, Gibberen, Gibberene
IUPAC Name 1,7-dimethyl-9H-fluorene
SMILES Cc1ccc2c(Cc3c(C)cccc23)c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C15H14/c1-10-6-7-13-12(8-10)9-15-11(2)4-3-5-14(13)15/h3-8H,9H2,1-2H3
Formula Weight 194.1096
Chemical Name or Material 1,7-Dimethylfluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.