missing translation for 'onlineSavingsMsg'
Learn More

1,8-Dihydroxyanthraquinone, TRC

Product Code. 30135094
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135094 10 g 10g
30138075 100 g 100g
17904865 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135094 Supplier Toronto Research Chemicals Supplier No. TRCD45015010G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-10-2
Molecular Formula C14 H8 O4
Molecular Weight (g/mol) 240.21
Synonym Anthraquinone, 1,8-dihydroxy- (8CI), 1,8-Dihydroxy-9,10-anthracenedione, 1,8-Dihydroxy-9,10-anthraquinone, 1,8-Dihydroxyanthraquinone, 1,8-Dioxyanthraquinone, Altan, Antrapurol, Chrysazin, Danthron, Danthrone, Dantron, Diaquone, Dionone, Dorbane, Istin, Istizin, Laxanorm, Laxanthreen, Laxipur, Laxipurin, Modane, NSC 38626, NSC 646568, NSC 7210, Zwitsalax
IUPAC Name 1,8-dihydroxyanthracene-9,10-dione
SMILES Oc1cccc2C(=O)c3cccc(O)c3C(=O)c12

Specifications

Quantity 10 g
InChI Formula InChI=1S/C14H8O4/c15-9-5-1-3-7-11(9)14(18)12-8(13(7)17)4-2-6-10(12)16/h1-6,15-16H
Formula Weight 240.0423
Chemical Name or Material Dantron
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.