missing translation for 'onlineSavingsMsg'
Learn More

1,8-Dinitropyrene (90%), TRC

Product Code. 30384369
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30384369 0.5 mg 0.5mg
30370540 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30384369 Supplier Toronto Research Chemicals Supplier No. TRCD4802700.5MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,8-Dinitropyrene
CAS 42397-65-9
Molecular Formula C16 H8 N2 O4
Quantity 0.5 mg
Synonym Pyrene, 1,8-dinitro-, 1,8-Dinitropyrene
InChI Formula InChI=1S/C16H8N2O4/c19-17(20)13-7-3-9-1-2-10-4-8-14(18(21)22)12-6-5-11(13)15(9)16(10)12/h1-8H
SMILES [O-][N+](=O)c1ccc2ccc3ccc(c4ccc1c2c34)[N+](=O)[O-]
IUPAC Name 1,8-dinitropyrene
Molecular Weight (g/mol) 292.25
Formula Weight 292.0484
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.